Would you like to log in again?
Tube and Cuvette Racks, Eppendorf®
Supplier: Eppendorf
0030119800 0030119819 0030119827 0030119851 0030119835 0030119860 0030119843
76181-440CS 69.68 USD InStock
76181-440 76181-450 76181-444 76338-494 76181-442 76181-448 76181-446
Tube and Cuvette Racks, Eppendorf®
Racks Tube Racks
Optimize workflow through user-friendly handling of clearly arranged samples
  • Stackable within one rack format and also between all rack variants; save laboratory space when not in use
  • Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Wavelike design; rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
  • Made of polypropylene (PP) for high chemical and physical resistance
  • Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement, without compromising handling
  • High flexibility allows different sized tubes to fit in the 1.5/2.0 mL, 5.0/15.0 mL, 5.0/15/50 mL, and cryogenic tube rack
  • Numbered tube positions support fast and easy orientation and error-free handling
  • Autoclavable (121°C for 20 minutes)
Benchtop racks are essential laboratory accessories for efficient processing, transport, and short-term storage of sample preparation tubes, cryogenic tubes, and cuvettes. The comprehensive Eppendorf® Tube Rack and Cuvette Rack system supports a clearly arranged and error-free handling of reaction vessels in manifold laboratory workflows. Optimized dimensions, design, and functionality result in various user benefits such as easy tube accessibility or flexible usage. These racks offer the optimal solution and perfect fit for all tubes and cuvettes commonly used in laboratories, with compact dimensions that require minimal space.
Eppendorf® Tube Racks and Cuvette Racks are ideal for sample processing, transport, and short- to mid-term storage of samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty