Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DBI-SALA® Python Safety™ Spill Control Buckets, Capital Safety

Supplier: ORS NASCO, INC.
DBI-SALA®
098-1500133
76111-184EA 188.29 USD InStock
76111-184
DBI-SALA® Python Safety™ Spill Control Buckets, Capital Safety
Buckets
  • Heavy-duty 24 oz. cotton duck canvas construction
  • Puncture resistant plating sewn into the base of the bucket
  • Six built-in connection points with a 10 lb. load rating for each
  • Convenient drawstring closure system
  • Features carabiner and hoisting strap

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty