Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DBI-SALA® ShockWave2™ Shock Absorbing Lanyards, ORS Nasco

Supplier: ORS Nasco
DBI-SALA®
098-1244448
76111-360EA 577.53 USD InStock
76111-360
DBI-SALA® ShockWave2™ Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Abrasion resistant tubular webbing for durability and longevity
  • Impact indicator allows user to visually inspect unit for a fall
  • Limits arresting forces to 900 lbs. or less
  • Reduces trip fall hazards, dragging, and snagging for complete freedom of movement
  • Twin-leg 100% tie-off style helps users remain connected at all times
  • Use with full body harnesses & appropriate anchorage connectors

Certifications: OSHA, stringent ANSI Z359.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty