Would you like to log in again?
Protecta PRO™ Stretch Shock Absorbing Lanyards, ORS Nasco
Supplier: ORS Nasco
Protecta PRO™
098-1340161
76111-398EA 170.81 USD InStock
76111-398
Protecta PRO™ Stretch Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Abrasion resistant polyester tubular webbing
  • User-friendly self-locking snap hook at center
  • Self-locking steel rebar hook at leg ends
  • Energy absorbing design limits fall arrest forces
Certifications: Meets all applicable industry standards including OSHA and ANSI Z359.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty