Would you like to log in again?
DBI-SALA® Rope Positioning Nylon Lanyards, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1232354
76111-358EA 154.8 USD InStock
76111-358
DBI-SALA® Rope Positioning Nylon Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Single-leg rope with snap hooks at each end
  • Long enough to be user friendly, but must be rigged in a way to eliminate free falls
  • Ideal for work positioning or restraint
  • Built-in RFID identification tag provides a way to record and store information on inspections
Rugged construction endures rough use, standing up to long periods of outdoor elements including sun exposure without tearing or fraying for added longevity. Each lanyard is hand spliced and dipped into a seizing solution for added durability. User-friendly self locking snap hooks at each end automatically lock and close for added safety, and allow opening and connection without getting thumbs or fingers in the way, even with gloves on.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty