Would you like to log in again?
Protecta Rebel™ Self Retracting Cable Lifelines, ORS Nasco
Supplier: ORS Nasco
Protecta® Rebel™
098-3590560
76111-508EA 1051.83 USD InStock
76111-508
Protecta Rebel™ Self Retracting Cable Lifelines, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Self-locking anchorage carabiner
  • Side lifeline exit with ergonomic handle
  • Rugged, lightweight thermoplastic housing
  • Stackable housing design
  • Integrated handle makes transport, set-up, use, and storage quick and easy
  • Abrasion-resistant and extremely durable lifeline provides added field longevity
  • Swiveling self-locking snap hook with impact indicator
  • Internal energy absorption system
  • Reserve lifeline system
Unique housing allows for stacking of multiple SRLs on top of each other to reduce overall space needed during storage. The retractable design provides freedom of movement, keeps the lifeline out of the worker's way, and reduces dragging, snagging, and trip falls. Speed sensing all metal brake system stops a falling worker within inches and provides smooth consistent operation, allowing the user to maintain normal work pace and not be interrupted with constant lock ups. Specialized internal absorption system maintains safe fall arrest forces for users and tools up to 420 lbs. (190 kg). Reserve lifeline system maintains a reserve for shock absorption when the entire lifeline is extended. Side lifeline exit with ergonomic handle enables smooth operation and reduced cable wear, and the handle provides added comfort and ease of use when making connections. Stainless steel and zinc plated steel hardware provides added corrosion resistance and extends the life of the device.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty