Would you like to log in again?
DBI-SALA® Advanced™ Confined Space Tripod Pulleys, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-8003238
76111-562EA 333.07 USD InStock
76111-562
DBI-SALA® Advanced™ Confined Space Tripod Pulleys, ORS Nasco
Fall Protection Fall Protection Kits
Confined space pulley bolts to 8000000 series tripod leg and routes winch/SRL lifeline up to 0.25" diameter. Loaded with features such as locking legs, simple detent pin style adjustment, quick-mount mechanical device bracket, safety chain to prevent movement, and safety shoes that incorporate a rubber sole for flat surfaces and spiked edges for slippery surfaces.
  • Easily bolts to tripod leg and provides a means to route mechanical device lifeline
  • Offers non-abrasive smooth operation and positions lifeline over the confined space entrance for efficient retrieval
  • Corrosion resistant, lightweight, and durable design will stand up to the elements and extend longevity
  • Detent pin style lifeline retention system allows user to quickly and easily route the mechanical device lifeline
  • Slim design hugs the tripod leg, staying out of the way and allowing user attachment
Versatile device fits most confined space entry/retrieval winches and 3-way self retracting lifeline cables and ropes for added flexibility. Extremely lightweight and portable, easily set-up by one worker, and can be transported from one location to another. Ideal for confined space entry and emergency rescue applications, the pulley can accommodate a variety of winches and fall arrest devices.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty