Would you like to log in again?
DBI-SALA® Advanced™ Aluminum Tripods, ORS Nasco
Supplier: ORS Nasco
098-8000000
76111-560EA 2145.69 USD InStock
76111-560
DBI-SALA® Advanced™ Aluminum Tripods, ORS Nasco
Fall Protection Fall Protection Kits
Lightweight and durable aluminum construction is easy to transport, set-up, and use. Tripods will stand up to the elements for extended longevity.
  • Built-in pulley system
  • Safety leg support chain
  • Quick connect winch/SRL mount bracket
  • Telescoping adjustable locking legs
  • Rubber safety shoes with spiked edges
  • Integrated 5,000 lbs. eyebolt anchor points
  • Equipped with i-Safe™ Integrated RFID identification tag
The built-in pulley system provides an easy way to route a mechanical devices lifeline into the confined space, and keeps it centered. Safety leg support chain prevents movement of the legs when the system is under load. Easily attach a mechanical device such as winch or 3-way self retracting lifeline directly to the tripod leg. Simple detent pin leg adjustment in 3" increments offers quick and easy set-up, allowing a custom fit. Rubber safety shoes with spiked edges offers a non-slipping design for both flat surfaces and uneven terrain. Integrated 5,000 lbs. eyebolt anchor points are ideal for attachment of secondary or back-up systems. Equipped with i-Safe™ Integrated RFID identification tag for recording and storing information on inspections and tracking safety equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty