Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA Lad-Saf™ X2 Detachable Cable Sleeves, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-6160030
76111-546EA 798.49 USD InStock
76111-546
DBI-SALA Lad-Saf™ X2 Detachable Cable Sleeves, ORS Nasco
Fall Protection Fall Arresters
For use with Lad-Saf™ Fixed Ladder Safety System.
  • Automatically follows the climber during ascent or descent
  • Two independent locking systems, including a mechanical and inertia locking system, in the event of a fall
  • Compact and lightweight design has stainless steel construction to stand up to harsh environments
  • 1,350 lb. breaking strength
  • 0.6875" opening size
One-handed attachment or removal from the cable delivers fast, easy, and safe connections. The sleeve can be attached anywhere along the cable for added versatility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty