Would you like to log in again?
DBI-SALA Roof Top Freestanding Counterweight Anchors, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-2100185
76111-194EA 5321.51 USD InStock
76111-194
DBI-SALA Roof Top Freestanding Counterweight Anchors, ORS Nasco
Fall Protection Fall Arresters
Freestanding system is approved for use on concrete, single ply and bitumen membrane, asphalt, and stone chippings for added jobsite flexibility. System does not attach to the working surface, reducing the possibility of damage. Built-in shock absorbing system for added safety.
  • Installs to flat or up to 5 degree sloped roofs
  • Non-penetrating design simply sits on working surface, reducing the possibility of damage
  • Energy absorbing design protects the worker and the structure in the event of a fall, limiting fall arrest forces to safe levels
  • Integral carrying handles for easy transport
  • Ideal for use with a shock absorbing lanyard, self retracting lifeline, or rope grab system for added versatility
  • Device can be removed and simply re-installed in a new location for added jobsite flexibility and lower overall costs
Supplied complete with sixteen 20 kg (45 lbs.) counterweights and four PVC coated base plates. Each countweight has an ergonomic carrying handle for easy set-up and user satisfaction.
Anchorage connectors are built from top quality materials guaranteeing high strength and durability to stand up to the toughest environments. Anchorage selection depends on the type of work being done.
Anchorages are sold by what they connect to (general/all purpose, steel, concrete, roof, or specialty), whether they are fixed or mobile, and whether they are permanent or portable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty