Would you like to log in again?
DBI-SALA Nano-Lok AF Aluminum Carabiners, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3101575
76111-444EA 516.29 USD InStock
76111-444
DBI-SALA Nano-Lok AF Aluminum Carabiners, ORS Nasco
Fall Protection Karabiners
  • Compact and lightweight design
  • Electric arc performance of 40 cal/cm2
  • 5,000 lb. breaking strength
  • The ultimate in arc flash, spark and fire resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty