Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA Nano-Lok AF Steel Carabiners with Snap Hook, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3101531
76111-436EA 486.37 USD InStock
76111-436
DBI-SALA Nano-Lok AF Steel Carabiners with Snap Hook, ORS Nasco
Fall Protection Karabiners
  • Compact and lightweight design
  • Electric arc performance of 40 cal/cm2
  • 5,000 lb. breaking strength
  • The ultimate in arc flash, spark and fire resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty