Would you like to log in again?
25 bp DNA Step Ladder, 100 µg, Promega
Supplier: Promega
G4511
PAG4511EA 188.47 USD InStock
PAG4511
25 bp DNA Step Ladder, 100 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Twelve DNA fragments ranging from 25bp to 300bp in 25bp increments includes a visible 1,800bp "backbone" fragment.
  • Twelve DNA Fragments of 25-300bp in 25bp Increments
  • Includes 1,800bp "backbone" fragment
  • Load 5microl/lane
  • Supplied at a concentration of 0.36µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 25bp DNA Step Ladder: Twelve DNA fragments ranging from 25bp to 300bp in 25bp increments. An 1,800bp backbone fragment is also visible. The 300bp band is ~3 times more intense than all other bands. Recommended loading: 5microl/lane.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty