Would you like to log in again?
Kanamycin Positive Control RNA, 1.2 kb, 5 µg, Promega
Supplier: Promega
C1381
PAC1381EA 105.02 USD InStock
PAC1381
Kanamycin Positive Control RNA, 1.2 kb, 5 µg, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
1.2 kb Kanamycin Positive Control RNA is supplied in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA.
  • Synthesis of double stranded cDNA from poly(A)+ RNA
Caution: Storage Requirements <−65 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty