Would you like to log in again?
Wizard SV 96 Genomic DNA Purification System, Promega
Supplier: Promega
Wizard®
A2370 A2371
PAA2370EA 301.26 USD InStock
PAA2370 PAA2371
Wizard SV 96 Genomic DNA Purification System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard SV 96 Genomic DNA Purification System provides a high-throughput, membrane-based technique for preparation of genomic DNA from cultured cells and tissue, including mouse tails.
  • 96-Well, Vacuum-Based DNA Purification From Cells or Tissues
  • Suitable for manual or high-throughput applications
  • DNA from mouse tails in 45-60 minutes; DNA from cells in 30 minutes
  • 20-30microg DNA per prep from 1.2cm mouse tail
The Wizard SV 96 Genomic DNA Purification System provides a high-throughput, membrane-based technique for consistent preparation of genomic DNA from cultured cells and tissue, including mouse tails. Amplifiable genomic DNA can be isolated from up to 5 x 10(6) cells, 20mg of tissue or up to 1.2cm of a mouse tail tip without a centrifugation clearing step. With the Wizard SV Genomic DNA purification system, genomic DNA is purified from cell lysates using 96-well vacuum filtration. Washing the bound DNA requires no disassembly of the manifold, and filtrate waste products are delivered directly to a vacuum trap, eliminating the need to empty waste collection trays. The Wizard SV Genomic DNA Purification System is designed for use either in a manual format or with Beckman Coulter or PerkinElmer automated instruments.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty