Would you like to log in again?
pTARGET Mammalian Expression Vector System, 20 reactions, Promega®
Supplier: Promega
A1410
PAA1410EA 538.58 USD InStock
PAA1410
pTARGET Mammalian Expression Vector System, 20 reactions, Promega®
Vectors, Plasmids and Libraries
Convenient system for cloning and expression of PCR products in mammalian cells.
  • Simple PCR Cloning for Mammalian Expression
  • Clone PCR products with T overhangs
  • CMV promoter for robust, consistent expression
  • Neomycin marker for selection of stable transfectants
The pTARGET Mammalian Expression Vector System is a convenient system for cloning PCR products and for expressing cloned PCR products in mammalian cells. The vector is prepared by digestion with EcoRV followed by addition of a 3' terminal thymidine to each end. These single 3'-T overhangs at the insertion site greatly improve the efficiency of ligation of a PCR product into the plasmid in two ways. First, the overhangs prevent recircularization of the vector; second, they provide a compatible overhang for PCR products generated by thermostable polymerases that add a single deoxyadenosine, in a template-independent fashion, to the 3'-ends of amplified fragments. The pTARGET Vector also contains a modified coding sequence of the alpha-peptide of beta-galactosidase, which allows recombinants to be selected using blue/white screening. The pTARGET Vector carries the human cytomegalovirus (CMV) immediate-early enhancer/promoter region to promote constitutive expression of cloned DNA inserts in mammalian cells. This vector also contains the neomycin phosphotransferase gene, a selectable marker for mammalian cells. The pTARGET Vector can be used for transient expression or stable expression by selecting transfected cells with the antibiotic G-418.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty