Would you like to log in again?
ADP-Glo Kinase Assay + CAMK2 gamma Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9201
PAV9201EA 1196.72 USD InStock
PAV9201
ADP-Glo Kinase Assay + CAMK2 gamma Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family.
  • Easily Screen and Profile CAMK2γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family, which are ubiquitous serine/threonine protein kinases that have been implicated in diverse effects of hormones and neurotransmitters. CAMK2 gamma has six alternatively spliced variants that encode six different isoforms. Some of these variants have been identified in human tumors. Transgenic mice expressing a partially calcium-independent mutant form of CAMKG showed 1.5- to 2-fold increase in the thymus of these mice, at least in part due to an increase in the life span of double-positive thymocytes. There was an increase in the number of T cells in the secondary lymphoid organs that had acquired an antigen-dependent memory phenotype. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CAMK2 gamma, 10ug (Human, recombinant; C-terminal truncation). MW: ~60kDa. Substrate: Autocamtide-2 (KKALRRQETVDAL-amide); derived from the autophosphorylation site (amino acid 283-290) on CaMKII. Other: Reaction Buffer, DTT, Ca2+/Calmodulin solution. CAMK2 gamma NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/818/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty