Would you like to log in again?
ADP-Glo Kinase Assay + LCK Kinase Enzyme System, 1 each, Promega
Supplier: Promega
V9491
PAV9491EA 1266.1 USD InStock
PAV9491
ADP-Glo Kinase Assay + LCK Kinase Enzyme System, 1 each, Promega
Assays Enzyme Assays
Recombinant full-length human LCK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LCK (p56lck) is a member of the src family of nonreceptor tyrosine kinases.
  • Easily Screen and Profile LCK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human LCK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LCK (p56lck) is a member of the src family of nonreceptor tyrosine kinases. It was identified as a gene rearranged and overexpressed in the murine lymphoma LSTRA, most likely as a result of the insertion of Moloney murine leukemia virus DNA immediately adjacent to the gene. LCK behaves as a proto-oncogene and can lead to cell transformation upon activation. A number of human cancer cell lines show overexpression of LCK, pointing to a possible role for this kinase in the initiation and maintenance of the transformed state in human cancers. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: LCK, 10ug (Human, recombinant full-length). MW: ~84kDa. Substrate: Poly (4:1 Glu, Tyr) Peptide. Other: Reaction Buffer, DTT, MnCl2. LCK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/3932/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty