Would you like to log in again?
Neutrophil/Monocyte Respiratory Burst Assay Kit, Cayman Chemical Company
Supplier: Cayman Chemical
601130-1
75817-102EA 499.84 USD InStock
75817-102
Neutrophil/Monocyte Respiratory Burst Assay Kit, Cayman Chemical Company
Assays Cellular Assays
Neutrophil/Monocyte Respiratory Burst Assay Kit provides PMA, dihydrorhodamine 123, and additional reagents necessary for inducing and quantifying a respiratory burst response in neutrophils and monocytes by flow cytometry. The assay can be performed on whole blood or on cells in various types of cell culture media. Because the assay reagents are not species-specific, this assay can be used in any species or cell type capable of producing a NADPH oxidase-dependent respiratory burst response.
  • Induce and quantify a respiratory burst response
  • Analyse by flow cytometry
Respiratory burst, or the rapid generation of reactive oxygen species from immune cells, is crucial for the destruction of invading microorganisms by phagocytes. During this process, professional phagocytes convert molecular oxygen to superoxide anions through the action of NADPH oxidase, which is then converted to reactive oxygen species with potent antimicrobial activity.
Delivery information: The kit includes dihydrorhodamine 123 assay reagent (50 µL), PMA (1 mM) assay reagent (50 µL), RBC lysis buffer (10X) (10 mL), bovine serum albumin assay reagent (5 g) and calcium chloride (1 M) assay reagent (1 mL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty