Would you like to log in again?
Fetal Bovine Serum, Heat Inactivated, Certified, U.S. Origin, Rockland™
Supplier: Rockland Immunochemicals
FBS-01-0500 FBS-01-0100
10802-770EA 226 USD InStock
10802-770 10802-772
Fetal Bovine Serum, Heat Inactivated, Certified, U.S. Origin, Rockland™
Sera (Serum)
Fetal bovine serum is used in a wide range of cell culture apllications.
  • Sterile bacteriphage
  • Store at −20 °C
This bovine serum is certified to originate in the United States as heat inactivated, performance, mycoplasma, virus, bacteriophage and endotoxin tested.
Caution: For in vitro diagnostic use only, not for human or animal therapeutic use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty