Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azodicarbonamide 97%

Supplier: Thermo Scientific Chemicals
Danger

AC10516-5000
103530-188EA 58.8 USD InStock
103530-188
Azodicarbonamide 97%
Azodicarbonamide


Formula: C₂H₄N₄O₂
MW: 116.08 g/mol
Melting Pt: 195…202 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007958
EINECS: 204-650-8
UN: 3242
ADR: 4.1,II
Order Now
Specification Test Results
Appearance Yellow to orange Powder
Infrared spectrum Conforms
Titration Iodometric ≥96.0 %
Water ≤0.2 %
Peroxide value ≤5 meq O2/kg

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty