Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
CIzyme Collagenase HA, Vitacyte
Supplier: Vitacyte
001-1000
101449-112EA 3461.54 USD InStock
101449-112
CIzyme Collagenase HA, Vitacyte
Microbiology Test Systems Microorganism Tests
CIzyme Collagenase HA is an aseptically prepared, lyophilized mixture of 60% purified Class I (C1) Collagenase and 40% purified Class II (C2) Collagenase from C. histolyticum
  • Manufactured for superior lot-to-lot enzyme consistency for improved performance consistency
  • Sufficient enzyme activity (>1,900 wunsch units) to release islets from average size adult human pancreata
  • Purification process eliminates the majority contaminating protease activity including clostripain (trypsin-like activity)
  • Low contaminating endotoxin levels (<10 EU/mg)
  • Lot specific certificate of analysis provided with complete QC analysis results
  • Highly purified collagenases for greater enzyme potency (>2.8 U/mg)
  • Lyophilized in a buffer containing calcium to ensure enzymes stability during initial reconstitution
CIzyme Collagenase HA is manufactured to provide the optimal collagenase activity for isolating cells from tissue or recovering adherent cells after in vitro culture and to isolate islets from human pancreata.
Delivery information: White lyophilized cake under vacuum in amber bottle sealed with butyl rubber stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty