Would you like to log in again?
Heat Sealing Foils, PCR Clean, Eppendorf
Supplier: Eppendorf
0030127854
75875-060CS 192.33 USD InStock
75875-060
Heat Sealing Foils, PCR Clean, Eppendorf
Seals Microplate Seals
The Eppendorf HeatSealer CS100 provides PCR plate with a tight seal, particularlty when heatsealing material is used.
  • Best protection against evaporation during PCR
  • Maximum adhesiveness at high temperatures
  • Adhesive flaps help to position the foil for sealing
  • Fluorescence applications, including real-time PCR
It provides optically clear films, especially the Masterclear® real-time PCR Film with its superior optical characteristics, are the ideal sealing material of choice for qPCR and other optical assays.
These sealing foils are hermetic sealing of multiwell plates, especially recommended for low reaction volumes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty