Would you like to log in again?
Aminopeptidase M
Supplier: EMD Millipore
164598-50U
80055-770EA 277.32 USD InStock
80055-770
Aminopeptidase M
Enzymes
Native amino peptidase M from porcine kidney. Metalloprotease that hydrolyzes N-terminal amino acids from almost all unsubstituted oligopeptides. It does not cleave X-Pro bonds or N-blocked amino acids.
  • White crystalline suspension
  • Useful in peptide sequencing
  • Inhibited by EDTA, bestatin and amastatin
Storage: 2 to 8 °C
Packaging: Packed in plastic ampoule.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty