Would you like to log in again?
Pepstatin A
Supplier: EMD Millipore
516481-5MG 516481-100MG 516481-5MG 516481-25MG 516481-25MG
80603-478EA 102.38 USD InStock
80603-478 CA80603-478 CA80603-476 80603-474 CA80603-474
Pepstatin A
Proteins and Peptides
Pepstatin A is a reversible inhibitor of aspartic proteases and inhibits cathepsin D, pepsin, and renin.
  • Reported to block apoptosis induced by inhibitor in PC12 cell lines
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty