Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: 186-2G9]

Supplier: Biotium

BNCB0178-500 BNCB0178-100
75915-042EA 547.44 USD InStock
75915-042 75915-040
Anti-ICAM3 Mouse Monoclonal Antibody (Biotin) [clone: 186-2G9]
Antibodies
This antibody recognizes the D1 domain of an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3 (Leucocyte Workshop VI). CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Clone 186-2G9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty