Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: C21.48A1]

Supplier: Biotium

BNC040433-100 BNC040433-500
75882-180EA 547.44 USD InStock
75882-180 75882-178
Anti-B2M Mouse Monoclonal Antibody (CF405S) [clone: C21.48A1]
Antibodies
The antibody recognizes the hidden determinant of β2-microglobulin (i.e. binding to its determinant is available only when the chain is separated from the HLA heavy chain. β2-microglobulin is a 12 kDa protein with a pI of 5.6. Serum beta2 microglobulin levels are a reflection of cell turnover. Levels rise with fever, inflammation, and infection. Increased serum levels are also seen in B-cell malignancies and in renal failure and may indicate a worse prognosis for patients with early-stage Hodgkins lymphoma. In urine, increased levels are seen in proximal renal tubular disease as well as renal transplant rejection. β2-microglobulin levels can rise either because its rate of synthesis has increased (e.g. in AIDS, malignant monoclonal plasma cell dyscrasia, solid tumours and autoimmune disease) or because of impaired renal filtration (e.g. due to renal insufficiency, graft rejection or nephrotoxicity induced by post-transplantation immunosuppressive therapy).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: Clone C21.48A1
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human, Non-human primates

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty