Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSF2 Rat Monoclonal Antibody (CF647) [clone: Clone BVD2-21C11]

Supplier: Biotium

BNC470662-100 BNC470662-500
75948-832EA 547.44 USD InStock
75948-832 75948-830
Anti-CSF2 Rat Monoclonal Antibody (CF647) [clone: Clone BVD2-21C11]
Antibodies
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: Clone BVD2-21C11
Conjugation: CF647
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty