Would you like to log in again?
BriskHeat® Samox High-Temperature Heating Tapes, Leads Same Ends, 120VAC, Cole-Parmer
Supplier: Cole Parmer
BriskHeat®
03106-11
76181-514EA 291.87 USD InStock
76181-514
BriskHeat® Samox High-Temperature Heating Tapes, Leads Same Ends, 120VAC, Cole-Parmer
Tapes
Get direct-contact heating on conductive surfaces.
  • Maximum exposure temperature of 1400ºF (760ºC)
  • Heavy-duty insulated design
These heating tapes feature a heavy-duty insulated design, for a maximum exposure temperature of 1400 ºF (760 ºC).
The 120 VAC tapes include a plug and the 240 VAC tapes are supplied with bare leads. The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty