Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Peptone from Gelatin, Millipore

Supplier: Millipore
1.07284.1000
EM1.07284.1000EA 264.6 USD InStock
EM1.07284.1000
Peptone from Gelatin, Millipore
Media for Microbiology Additives and Supplements for Microbiology
Gelatin peptone is a pancreatic digest of gelatin. It is suitable for preparing media for organisms.

  • Appearance: Lightly yellow to lightly brownishyellow fine powder
  • Storage: 15° C to +25° C
  • Bulk Density: 420 kg/m3
  • Solubility: 550 g/l

Gelatin peptone is used in cell cultures, bacterial fermentation requiring high hydroxyproline content, low carbohydrates, cystine
and tryptophan levels and the cultivation of non fastidious microorganisms.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty