Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Accessories for OpalMist™ and PolyCon Nebulizers in ICP-OES and ICP-MS Instruments, PerkinElmer
Supplier: PerkinElmer
N0775358 N0775358
97013-598EA 1214.74 USD InStock
97013-598 CA97013-598
Accessories for OpalMist™ and PolyCon Nebulizers in ICP-OES and ICP-MS Instruments, PerkinElmer
Atomizers
This Eluo Nebulizer Cleaner is for use with old style OpalMist™ and PolyCon Nebulizers in ICP-OES and ICP-MS instruments. The Eluo is designated to efficiently deliver a cleanser through the nebulizer capillary to dislodge particle build-up and thoroughly clean the nebulizer. Use the Eluo regularly to maintain performance and prolong the life of your nebulizer.
Eluo Nebulizer Cleaner Particle build-up in a nebulizer capillary and tip causes sample flow to be constricted, thereby reducing nebulizer efficiency and performance. Previously there was no simple way to effectively clean and unblock nebulizers. Now, blocked nebulizers can be safely and easily restored to optimum performance with a revolutionary new cleaning instrument - the Eluo.
Cleaning your nebulizer is now a simple three step process: Fill your Eluo with Methanol. Insert your nebulizer, tip first, into the Eluo. Clean your nebulizer by pressing the Plunger down. Use the Eluo regularly to maintain nebulizer performance and prolong nebulizer life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty