Would you like to log in again?
DuPont™ Tychem® 5000 Bib Coveralls
Supplier: DuPont
Tychem®
C3360TTN4X000600 C3360TTN3X000600 C3360TTNMD000600 C3360TTNXL000600 C3360TTNSM000600 C3360TTN2X000600 C3360TTNLG000600
76187-330CS 319.28 USD InStock
76187-330 76187-332 76187-334 76187-412 76187-336 76187-414 76187-416
DuPont™ Tychem® 5000 Bib Coveralls
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Bib style
  • Open ankles
  • Adjustable webbing straps with buckle closure
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These coveralls are tan.
These overalls are bib-style with adjustable wedding straps. They are typically worn in combination with the C3670T jacket or in applications were potential exposure is below the waist.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty