Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium iron (II) sulfate 0.1 N

Supplier: AQUA SOLUTIONS MS

3840-1L 3840-500ML 3840-100ML 3840-4L
101225-968EA 31.23 USD InStock
101225-968 101226-914 101224-260 101226-510
Ammonium iron (II) sulfate 0.1 N
Haematoxylin Harris
Certifications: Traceable to NIST.


Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Density: 1.04213 g/mL
MDL Number: MFCD00150530
CAS Number: 7783-85-9
UN: 1760
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty