Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Cyclodextrine ≥98% (by HPLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma
Synonyms: Schardinger β-Dextrinβ-CyclodextrinBetadexCycloheptaamyloseCyclomaltoheptaoseCaraway

239760-25GM 239760-500GM 239760-500GM
80503-696EA 920.63 USD InStock
CA80503-696 80503-696 80503-694
β-Cyclodextrine ≥98% (by HPLC), Calbiochem®, Millipore®
β-Cyclodextrine
β-Cyclodextrin are homogeneous cyclic α1,4-linked D-glucopyranose units in a seven-member ring which forms clathrates. Suitable for use with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins.


Formula: C₄₂H₇₀O₃₅
MW: 1134.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty