Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Colophony (Rosin gum) working solution 10%

Supplier: Electron Microscopy Sciences
Synonyms: Rosin, gumGum rosinRosin

20550-15
101410-784EA 58.34 USD InStock
101410-784
Colophony (Rosin gum) working solution 10%
Colophony (Rosin gum)
Rosin Alcohol Solution
Rosin is available in alcohol stock and working solutions.


MDL Number: MFCD00132205
CAS Number: 8050-09-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty