Would you like to log in again?
Elution Buffer, Promega
Supplier: Promega
MD1421
PAMD1421EA 13.72 USD InStock
PAMD1421
Elution Buffer, Promega
Buffers Electrophoresis Buffers Running Buffers
Elution Buffer, Blood, is a component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370) and Maxwell 16 Blood DNA Purification Kit (Cat.# AS1010).
  • Buffer Used With Automated Blood DNA Purification Systems
  • Component of the MagneSil(R) Blood Genomic, Max Yield System (Cat.# MD1360), MagneSil(R) ONE, Fixed Yield Blood Genomic System (Cat.# MD1370) and Maxwell(R) 16 Blood DNA Purification Kit (Cat.# AS1010)
Elution Buffer, Blood, is a component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370) and Maxwell 16 Blood DNA Purification Kit (Cat.# AS1010).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty