Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cefonicid sodium 832 - 970 mcg/mg (by HPLC)

Supplier: CHEM IMPEX INTL INC MS
Synonyms: MonocidDisodium (6R,7R)-7-[[(2R)-2-hydroxy-1-oxo-2-phenylethyl]amino]-8-oxo-3-[[[1-(sulfonatomethyl)-5-tetrazolyl]thio]methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylateDisodium (6R,7R)-7-[[(2R)-2-hydroxy-1-oxo-2-phenylethyl]amino]-8-oxo-3-[[[1-(sulphonatomethyl)-5-tetrazolyl]thio]methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

15160-5G
102633-088EA 131.74 USD InStock
102633-088
Cefonicid sodium 832 - 970 mcg/mg (by HPLC)
Cefonicid sodium


Formula: C₁₈H₁₆N₆Na₂O₈S₃
MW: 586.51 g/mol
Melting Pt: >160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00941437
CAS Number: 61270-78-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty