Would you like to log in again?
Opal™ Reagent Pack, Perkinelmer
Supplier: Perkin Elmer
FP1494001KT FP1495001KT FP1497001KT FP1487001KT FP1496001KT FP1488001KT
MSPP-FP1494001EA 346.15 USD InStock
MSPP-FP1494001 MSPP-FP1495001 MSPP-FP1496001 MSPP-FP1487001 MSPP-FP1488001 MSPP-FP1497001
Opal™ Reagent Pack, Perkinelmer
Stains Histology Stains
Opal™ is a practical workflow for simultaneous detection of up to 6 tissue biomarkers plus nuclear counterstain, within a single image. The method is similar to standard immunohistochemistry and is accessible to many laboratories where standard IHC method development is performed.
Users should source primary antibodies and other workflow reagents separately. Before each procedure, dilute reconstituted Opal Fluorophore in 1X Amplification Diluent (not included) to make Opal fluorophore working solution.
Pack contains enough reagent to stain 50-slides with the following dilution ratios:
1:100 for manual kits
1:150 for automated kits
Store dry reagent in the dark at –20 °C. Upon reconstituting in DMSO, store in the dark at 4 °C.
Delivery information: The Opal reagent pack contains opal reagent, 1 vial, Dimethyl sulfoxide (DMSO), 1 x 100 µl.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty