Would you like to log in again?
Whatman™ MembraClear Cellulose Ester Membrane Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
7141-047 7141-047 7141-025
76200-228PK 298.3 USD InStock
76200-228 CA76200-226 CA76200-228
Whatman™ MembraClear Cellulose Ester Membrane Filters, Whatman products (Cytiva)
Filters Membrane Filters
These mixed cellulose ester membrane filters are designed for asbestos sampling using the membrane filter method for phase contrast microscopy. Asbestos sampling isolates these fibers from circulating air to determine concentrations.
  • Designed for use with the 'hot block method'
  • White with black grid for high contrast during asbestos testing using microscopy
Becomes transparent with acetone vapor treatment, distinguishing fibers from crystalline artifacts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty