Would you like to log in again?
Odyssey® One-Color Protein Molecular Weight Marker, LI-COR®
Supplier: LI-COR®
928-40000
102673-496EA 286.77 USD InStock
102673-496
Odyssey® One-Color Protein Molecular Weight Marker, LI-COR®
Electrophoresis Reagents Molecular Weight Markers
The Odyssey One-Color Protein MW Marker provides a ladder of convenient, consistent sizes for both visible and 700 channel near-infrared detection.
  • Includes ten protein bands from 10 to 250 kDa
  • Three reference bands at 25, 50, and 75 kDa are three times as intense as other bands
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty