Would you like to log in again?
Aurora A Kinase Enzyme System, Promega
Supplier: Promega
V1931 V1932
PAV1931EA 612.3 USD InStock
PAV1931 PAV1932
Aurora A Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant full-length human Aurora A was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora A belongs to a multigenic family of mitotic serine/threonine kinases, which are involved in the control of chromosome segregation.
  • Easily Screen and Profile Aurora A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant full-length human Aurora A was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora A belongs to a multigenic family of mitotic serine/threonine kinases, which are involved in the control of chromosome segregation. Aurora A is involved in centrosome separation, duplication and maturation as well as in bipolar spindle assembly and stability. Aurora A is expressed and active at the highest level during G2-M phase of the cell cycle. Overexpression of Aurora A has been found to be correlated with the grade of various human solid tumors. Ectopic Aurora A overexpression in any culture cell line leads to polyploidy and centrosome amplification. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: Aurora A, 10ug (Human, recombinant full-length). MW: ~72kDa. Substrate: Native Swine Myelin Basic Protein (MBP). Other: Reaction Buffer, DTT. Aurora A NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/6790/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty