Would you like to log in again?
Bacterial Strain NM522, Promega
Supplier: Promega
P2301
PAP2301EA 61.59 USD InStock
PAP2301
Bacterial Strain NM522, Promega
Nucleic Acid Reagents Cloning Kits and Ligation
NM522 contains an F' episome, which is required for production of ssDNA and for blue/white color selection.
  • Produces ssDNA with F´ Episome
  • Can be used for blue/white color selection when screening colonies
NM522 contains an F' episome, which is required for production of ssDNA and for blue/white color selection. To maintain the F', NM522 should be grown on minimal (M-9) medium. Genotype: supE, thi, Delta(lac-proAB), hsd5 (r-, m-), [F', proAB, lacI(q)ZDeltaM15].
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty