Would you like to log in again?
Bovine Serum Albumin (BSA), Acetylated
Supplier: Promega
R9461 R3961
PAR3961EA 47.67 USD InStock
PAR3961 PAR9461
Bovine Serum Albumin (BSA), Acetylated
Proteins and Peptides
Used as an enzyme stabilizer or as a carrier protein. Acetylated to be nuclease-free.
  • Quality tested and certified free of DNase and RNase activity
Bovine Serum Albumin, Acetylated, can be used as an enzyme stabilizer or as a carrier protein. It is prepared by a modification of the method of Gonzalez et al. and dialyzed extensively with deionized water to remove impurities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty