Would you like to log in again?
Broad Range Protein Molecular Weight Markers, 100 lanes, Promega
Supplier: Promega
V8491
PAV8491EA 181.05 USD InStock
PAV8491
Broad Range Protein Molecular Weight Markers, 100 lanes, Promega
Electrophoresis Reagents Molecular Weight Markers
10-225kDa; 9 bands; ready to load. Protein staining required for detection.
  • Nine Evenly Spaced Bands of 10-225kDa for Use with SDS-PAGE
  • 50kDa band is 3X intensity for use as a reference
  • Ready-to-load markers
  • Supplied at a concentration of 5microl/lane
The Broad Range Protein Molecular Weight Markers consist of nine clearly identifiable bands at convenient molecular weights. The protein sizes are 10, 15, 25, 35, 50, 75, 100, 150 and 225kDa. Each protein is present at a concentration of 0.1ug/ul, except for the 50kDa protein, which is present at 0.3ug/ul and serves as a reference indicator, having triple the intensity of the other proteins. All other proteins appear with equal intensity on the gel. These markers are intended for use as a size standard when performing SDS-PAGE (sodium dodecyl sulfate-polyacrylamide gel electrophoresis) for estimation of the molecular weight of the protein of interest. Note that they are not stained.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty