Would you like to log in again?
cAMP-Dependent Protein Kinase
Supplier: Promega
V5161
PAV5161EA 405.5 USD InStock
PAV5161
cAMP-Dependent Protein Kinase
Enzymes
cAMP-Dependent Protein Kinase (PKA), Catalytic Subunit, may be used to phosphorylate target proteins or for in vitro enzymological studies.
  • Purified Kinase for Protein Phosphorylation and Signal Transduction Studies
  • Purified from recombinant E. coli expressing the catalytic subunit of bovine PKA; 90% pure
  • Supplied at a concentration of 1.5-3mg/ml
The purified 40kDa cAMP-Dependent Protein Kinase (PKA), Catalytic Subunit, may be used to phosphorylate target proteins or for in vitro enzymological studies of neural and hormonal signal transduction. Intracellular targets include ion channels, transcriptional activator proteins, and regulatory enzymes of glycogen metabolism.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty