Would you like to log in again?
CK1 gamma 1 Kinase Enzyme System, Promega
Supplier: Promega
V6578 V4100
PAV4100EA 612.3 USD InStock
PAV4100 PAV6578
CK1 gamma 1 Kinase Enzyme System, Promega
Assays Enzyme Assays
Recombinant human CK1 gamma 1 (21-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Easily Screen and Profile CK1γ1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human CK1 gamma 1 (21-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. CK1γ1 or casein kinase 1, gamma 1 is one of the most abundant serine/threonine kinase in eukaryotic cells and is mainly involved in growth and morphogenesis. CK1γ1 possesses the C-terminal sequence motif (MTM), which it shares with CSNK1G2 and CSNK1G3; this motif is associated with heterologous carboxy-terminal sequences. CK1γ1 couples Wnt receptor activation to the cytoplasmic signal transduction apparatus. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: CK1γ1, 10microg (Human, recombinant; amino acids 21-end). MW: ~70-76kDa. Substrate: Native Casein Protein; purified from bovine milk. Other: Reaction Buffer, DTT. CK1γ1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/53944/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty