Would you like to log in again?
Diamond™ Nucleic Acid Dye
Supplier: Promega
Diamond™
H1181
PAH1181EA 99.98 USD InStock
PAH1181
Diamond™ Nucleic Acid Dye
Stains Electrophoresis Stains
Diamond Nucleic Acid Dye is a sensitive fluorescent dye that binds to single-stranded DNA, double-stranded DNA and RNA, and can be used to stain and visualize nucleic acids in gels.
  • Detect and visualise small amounts of nucleic acids after gel electrophoresis
  • Store at room temperature for quick and easy use without thawing
  • Use with a variety of common gel types and imaging equipment
  • Concentrated dye stable for up to 90 days at room temperature
Diamond Nucleic Acid Dye is a sensitive fluorescent dye that binds to single-stranded DNA, double-stranded DNA and RNA, and can be used to stain and visualize nucleic acids in gels. Diamond Nucleic Acid Dye is compatible with denaturing and native agarose and polyacrylamide gels and can be imaged with any standard imaging system, such as by UV transillumination with a Polaroid or digital camera, GE ImageQuant or Bio-Rad Gel Doc systems. The concentrated dye is stable for up to 90 days at room temperature. Diamond Nucleic Acid Dye does not require prewashing or destaining of gels. It is more much more sensitive than ethidium bromide, so less sample nucleic acid and nucleic acid markers are required for visualization, resulting in increased savings with every gel you run.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty