Would you like to log in again?
DNA-Dependent Protein Kinase Substrate Peptide
Supplier: Promega
V5671
PAV5671EA 353.9 USD InStock
PAV5671
DNA-Dependent Protein Kinase Substrate Peptide
Proteins and Peptides
DNA-Dependent Protein Kinase Peptide Substrate is a highly specific substrate for DNA-PK, with the sequence Glu-Pro-Pro-Leu-Ser-Gln-Glu-Ala-Phe-Ala-Asp-Leu-Trp-Lys-Lys.
  • Highly Specific Substrate for DNA-PK
  • Supplied at 10mg/ml in water
  • Km value for peptide is 760microM
DNA-Dependent Protein Kinase (DNA-PK) Peptide Substrate is a highly specific substrate for DNA-PK, with the sequence Glu-Pro-Pro-Leu-Ser-Gln-Glu-Ala-Phe-Ala-Asp-Leu-Trp-Lys-Lys. The K(m) value for this peptide is 760uM. DNA-PK is a nuclear serine/threonine protein kinase that, when activated by DNA, phosphorylates several DNA-binding substrates, including the tumor suppressor protein p53, the simian virus 40 (SV40) large T antigen and several transcription factors (e.g., SP1, OCT-1, c-Fos and serum response factor).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty