Would you like to log in again?
DNA-Dependent Protein Kinase
Supplier: Promega
V5811
PAV5811EA 356.29 USD InStock
PAV5811
DNA-Dependent Protein Kinase
Enzymes
DNA-Dependent Protein Kinase (DNA-PK) phosphorylates several DNA-binding substrates in vitro, including the tumor suppressor protein p53, the SV40 large T antigen and several transcription factors.
  • Purified DNA-PK for DNA-Binding Protein Phosphorylation Studies
  • Phosphorylates several DNA-binding proteins in vitro, including p53, SV40 large T antigen and several transcription factors.
DNA-Dependent Protein Kinase (DNA-PK) phosphorylates several DNA-binding substrates in vitro, including the tumor suppressor protein p53, the SV40 large T antigen and several transcription factors. DNA-PK is thought to play a role in controlling gene regulation and cell growth. DNA-PK is isolated from HeLa nuclear extracts as a complex consisting of a 400kDa catalytic subunit and a 155kDa heterodimeric DNA-binding component named Ku, which itself consists of subunits of approximately 85kDa and 70kDa.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty