Would you like to log in again?
DNA-PK Kinase Enzyme System, 2,500 units, Promega
Supplier: Promega
V4106
PAV4106EA 647.81 USD InStock
PAV4106
DNA-PK Kinase Enzyme System, 2,500 units, Promega
Assays Enzyme Assays
Native human DNA-Dependent Protein Kinase purified from HeLa cells. Consists of a ~460kDa catalytic subunit and a heterodimeric DNA-binding subunit (Ku) containing an 85kDa and a 70kDa peptide (1). Gene sequence can be found at accession number NM_006904.
  • Easily Screen and Profile DNA-PK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Native human DNA-Dependent Protein Kinase (DNA-PK) purified from HeLa cells. DNA-PK consists of an approximately 460kDa catalytic subunit and a heterodimeric DNA-binding subunit (Ku) containing an 85kDa and an 70kDa peptide (1). The gene sequence can be found at accession number NM_006904. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: DNA-PK, 2,500 units (Human, native full-length). MW: ~460kDa (catalytic subunit); ~85kDa (Ku subunit 1); ~70kDa (Ku subunit 2). Substrate: DNA-Dependent Protein Kinase Peptide Substrate (EPPLSQEAFADLWKK). Other: Reaction Buffer, DNA-PK Activation Buffer, DTT. DNA-PK NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/5591/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty